Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELMO1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986820
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6820 Supplier Bioss Supplier No. BS5807R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. Similarity to a C. elegans protein suggests that this protein may function in apoptosis and in cell migration. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.

Specifications

Antigen ELMO1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ELMO1
Gene Accession No. Q92556
Gene Alias 6330578D22Rik; BOS_4263; C230095H21Rik; CED12; CED-12; ced-12 homolog 1; Elmo1; ELMO-1; ELMO1s; engulfment and cell motility 1; engulfment and cell motility 1 (ced-12 homolog, C. elegans); engulfment and cell motility 1 splice 1; engulfment and cell motility 1, ced-12 homolog; Engulfment and cell motility protein 1; im:7151147; KIAA0281; MGC126406; OTTHUMP00000207056; OTTHUMP00000207060; Protein ced-12 homolog; wu:fq24e08; zgc:85612
Gene Symbols ELMO1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ELMO.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9844
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.