Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELMO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582636
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582636 Supplier Invitrogen™ Supplier No. PA582636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CDGWSLPNPE YYTLRYADGP QLYITEQTRS DIKNGTILQL AISPSRAARQ LMERTQSSNM ETRLDAMKEL AKLSAD Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 97%.

The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein. Similarity to a C. elegans protein suggests that this protein may function in phagocytosis of apoptotic cells and in cell migration. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMO2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELMO2
Gene Accession No. Q96JJ3
Gene Alias 1190002F24Rik; CED12; CED-12; ced-12 homolog 2; CED-12 homolog A; CED12A; Ced-12A; Elmo2; ELMO-2; engulfment and cell motility 2; engulfment and cell motility 2, ced-12 homolog; engulfment and cell motility protein 2; FLJ11656; hCed-12A; KIAA1834; PH domain protein CED12A; protein ced-12 homolog A; RP11-394O2.2
Gene Symbols ELMO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELMO2. Recombinant protein control fragment (Product #RP-92080).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63916
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.