Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELMO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113177
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113177 Supplier Invitrogen™ Supplier No. PA5113177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ELMO2 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein. Similarity to a C. elegans protein suggests that this protein may function in phagocytosis of apoptotic cells and in cell migration. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMO2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ELMO2
Gene Accession No. Q8BHL5, Q96JJ3
Gene Alias 1190002F24Rik; CED12; CED-12; ced-12 homolog 2; CED-12 homolog A; CED12A; Ced-12A; Elmo2; ELMO-2; engulfment and cell motility 2; engulfment and cell motility 2, ced-12 homolog; engulfment and cell motility protein 2; FLJ11656; hCed-12A; KIAA1834; PH domain protein CED12A; protein ced-12 homolog A; RP11-394O2.2
Gene Symbols ELMO2
Host Species Rabbit
Immunogen Recombinant Human Engulfment and cell motility protein 2 protein (304-371AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140579, 63916
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.