Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELMO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554423 Supplier Invitrogen™ Supplier No. PA554423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVKSEVPLDR LLVHLQVMNQ QLQTKAMALL TALLQGASPV ERKHMLDYLW QRNLRQFIYK NIIHSAAPMG DEMAHHLYVL QALMLGL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

The protein encoded by this gene is similar to a C. elegans protein that functions in phagocytosis of apoptotic cells and in cell migration. Other members of this small family of engulfment and cell motility (ELMO) proteins have been shown to interact with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMO3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELMO3
Gene Accession No. Q96BJ8
Gene Alias 9930107J06; BC058752; Ced12; CED-12; ced-12 homolog 3; Elmo3; ELMO-3; engulfment and cell motility 3; engulfment and cell motility 3, ced-12 homolog; engulfment and cell motility protein 3; FLJ13824
Gene Symbols ELMO3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELMO3. Recombinant protein control fragment (Product #RP-92094).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.