Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ELMOD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | ELMOD1 |
---|---|
Concentration | 0.3 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111099
|
Invitrogen™
PA5111099 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111100
|
Invitrogen™
PA5111100 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: THFYNIAPEA PTLSHFQQTF CYLMHEFHKF WIEEDPMDIM EFNRVREKFR KRIIKQLQNP DMALCPHFAA SEGLINM.
Acts as a GTPase-activating protein (GAP) toward guanine nucleotide exchange factors like ARL2, ARL3, ARF1 and ARF6, but not for GTPases outside the Arf family.Specifications
ELMOD1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
ELMOD1 | |
4831417L10; AI428506; AI834991; ELMO domain containing 1; ELMO domain-containing protein 1; ELMO/CED-12 domain containing 1; Elmod1; RGD1564186 | |
ELMOD1 | |
Primary | |
Antigen affinity chromatography |
0.3 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N336 | |
55531 | |
Recombinant Protein corresonding to Human ELMOD1. Recombinant protein control fragment (Product #RP-97309). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title