Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELMOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111100
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111100 100 μL
PIPA5111099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111100 Supplier Invitrogen™ Supplier No. PA5111100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRKLTGRCEL QRICYNTKPG ASRTMKIETS LRDSKSKLLQ TSVSVHPDAI EKTIEDIMEL KKINPDVNPQ LGISLQACLL QIVGYRN Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

Acts as a GTPase-activating protein (GAP) toward guanine nucleotide exchange factors like ARL2, ARL3, ARF1 and ARF6, but not for GTPases outside the Arf family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMOD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELMOD1
Gene Accession No. Q8N336
Gene Alias 4831417L10; AI428506; AI834991; ELMO domain containing 1; ELMO domain-containing protein 1; ELMO/CED-12 domain containing 1; Elmod1; RGD1564186
Gene Symbols ELMOD1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ELMOD1. Recombinant protein control fragment (Product #RP-97308).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.