Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELMOD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589885 Supplier Invitrogen™ Supplier No. PA589885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWEFFYGHFF RFWMKWLLRQ MTGKCELQRI FDTYVGAQRT HRIENSLTYS KNKVLQKATH VVQSEVDKYV DDIMKEKNIN PEKDASFKIC M; Positive Samples: Rat lung.

This gene encodes one of six engulfment and motility (ELMO) domain-containing proteins. This gene is thought to play a role in antiviral responses. Mutations in this gene may be involved in the cause of familial idiopathic pulmonary fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMOD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ELMOD2
Gene Alias 9830169G11Rik; ELMO domain containing 2; ELMO domain-containing protein 2; ELMO/CED-12 domain containing 2; Elmod2
Gene Symbols ELMOD2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 5-95 of human ELMOD2 (NP_7149131).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 688581
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.