Learn More
Invitrogen™ ELMOD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109683
Description
Immunogen sequence: QMTGKCELQR IFDTYVGAQR THRIENSLTY SKNKVLQKAT HVVQSEVDKY VDDIMKEKNI NPEKDASFKI CMKMCLLQIT GYKQLYLDVE SVRKRPYDSD NLQHEELLMK LWNLLMPTKK LNARISKQWA EIGFQGDDPK TDFRGMGILG LINLVYFSEN YTSEAHQILS RSNHPKLGYS YAIVGINLTE MAYSLLKSEA LKFHLYNLVP GIPTMEHFHQ FYCYLVYEFD KFWFEEEPES IMYFNLYREK FHEKIKGLLL DCNVALTLKV.
This gene encodes one of six engulfment and motility (ELMO) domain-containing proteins. This gene is thought to play a role in antiviral responses. Mutations in this gene may be involved in the cause of familial idiopathic pulmonary fibrosis.
Specifications
| ELMOD2 | |
| Polyclonal | |
| Unconjugated | |
| ELMOD2 | |
| 9830169G11Rik; ELMO domain containing 2; ELMO domain-containing protein 2; ELMO/CED-12 domain containing 2; Elmod2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 244548 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BGF6 | |
| ELMOD2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-293 of human ELMOD2 (NP_714913.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.