Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELMOD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596711 Supplier Invitrogen™ Supplier No. PA596711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATFLHLAHVW RTQRKTISDS GFVLKELEVL AKKSPRRLLK TLELYLARVS KGQASLLGAQ KCYGPEAPPF KDLTFTGESD LQSHSSEGVW LI; Positive Samples: A-549, Mouse brain, Rat brain; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, kinocilium, stereocilium.

RBM29 (RNA binding motif protein 29), also known as ELMOD3 (ELMO domain-containing protein 3) or RBED1 (RNA-binding motif and ELMO domain-containing protein 1), is a 381 amino acid protein that contains one ELMO domain and exists as six alternatively spliced isoforms. The gene encoding RBM29 maps to human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMOD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ELMOD3
Gene Accession No. Q5XIQ2, Q91YP6, Q96FG2
Gene Alias AI844780; C330008I15Rik; deafness, autosomal recessive 88; DFNB88; ELMO domain containing 3; ELMO domain-containing protein 3; ELMO/CED-12 domain containing 3; Elmod3; Kcmf1; liver-specific organic anion transporter 3TM12; LST3; organic anion transporter LST-3b; potassium channel modulatory factor 1; PP4068; Rbed1; RBM29; RNA binding motif and ELMO domain 1; RNA binding motif and ELMO/CED-12 domain 1; RNA binding motif protein 29; RNA-binding motif and ELMO domain-containing protein 1; RNA-binding motif protein 29; RNA-binding protein 29
Gene Symbols ELMOD3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 290-381 of human ELMOD3 (NP_0011284931).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232089, 297342, 84173
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.