Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELMSAN1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171912AF350
Description
ELMSAN1 Polyclonal antibody specifically detects ELMSAN1 in Human samples. It is validated for Western BlotSpecifications
ELMSAN1 | |
Polyclonal | |
Western Blot | |
c14_5541, C14orf117, C14orf43, chromosome 14 open reading frame 117, chromosome 14 open reading frame 43, ELM2 and Myb/SANT-like domain containing 1, ELMSAN1, FLJ00335, hypothetical protein LOC91748, LSR68 | |
The immunogen for this product maps to a region between residue 550 and 600 of human ELMSAN1 Open Reading Frame 43 using the numbering given in entry NP_001036783.1 (GeneID 91748). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
91748 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction