Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELMSAN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171912
Description
ELMSAN1 Polyclonal specifically detects ELMSAN1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Knockdown Validated.Specifications
ELMSAN1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
c14_5541, C14orf117, C14orf43, chromosome 14 open reading frame 117, chromosome 14 open reading frame 43, ELM2 and Myb/SANT-like domain containing 1, ELMSAN1, FLJ00335, hypothetical protein LOC91748, LSR68 | |
Rabbit | |
Affinity Purified | |
RUO | |
91748 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
NP_001036783.1 | |
ELMSAN1 | |
The immunogen for this product maps to a region between residue 550 and 600 of human ELMSAN1 Open Reading Frame 43 using the numbering given in entry NP_001036783.1 (GeneID 91748). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction