Learn More
Invitrogen™ ELMSAN1 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 85%; Pig: 92%; Rabbit: 85%; Rat: 93%.
Specifications
Specifications
| Antigen | ELMSAN1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ELMSAN1 |
| Gene Accession No. | Q6PJG2 |
| Gene Alias | 9430029N19Rik; C130039O16Rik; c14_5541; C14orf117; C14orf43; C8H14orf43; ELM2 and Myb/SANT domain containing 1; ELM2 and Myb/SANT-like domain containing 1; ELM2 and SANT domain-containing protein 1; ELMSAN1; Gm260; LOW QUALITY PROTEIN: ELM2 and SANT domain-containing protein 1; LSR68; MIDEAS; mitotic deacetylase associated SANT domain protein; RGD1306119 |
| Gene Symbols | ELMSAN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.