Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELMSAN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551825 Supplier Invitrogen™ Supplier No. PA551825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQEEVEVDIK ASDILILRSH ESNAPGSAGG QASEKPREGT GKSRRALPFS EKKKKTETFS KTQNQENTFP CK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 71%.

ELMSAN1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleus; DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMSAN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELMSAN1
Gene Accession No. Q6PJG2
Gene Alias 9430029N19Rik; C130039O16Rik; c14_5541; C14orf117; C14orf43; C8H14orf43; ELM2 and Myb/SANT domain containing 1; ELM2 and Myb/SANT-like domain containing 1; ELM2 and SANT domain-containing protein 1; ELMSAN1; Gm260; LOW QUALITY PROTEIN: ELM2 and SANT domain-containing protein 1; LSR68; MIDEAS; mitotic deacetylase associated SANT domain protein; RGD1306119
Gene Symbols ELMSAN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELMSAN1. Recombinant protein control fragment (Product #RP-89098).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91748
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.