Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELN Mouse anti-Human, Unconjugated, Clone: ELN/1981, Abnova™

Catalog No. 89337674 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-674 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-674 Supplier Abnova Corporation Supplier No. MAB21181
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ELN Monoclonal antibody specifically detects ELN in Human samples. It is validated for IHC-P.

Specifications

Antigen ELN
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ELN/1981
Conjugate Unconjugated
Description elastin
Gene ELN
Gene Alias FLJ38671, FLJ43523, SVAS, WBS, WS
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human ELN
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2006
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.