Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5115175

Description
Antibody detects endogenous levels of total Elongin A1.
This gene encodes the transcriptionally active subunit of the SIII (or elongin) transcription elongation factor complex, which also includes two regulatory subunits, elongins B and C. This complex acts to increase the rate of RNA chain elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites along the DNA template. Whereas a related protein with similar function, elongin A, is ubiquitously expressed, the encoded protein is specifically expressed in the testis, suggesting it may have a role in spermatogenesis.
Specifications
ELOA | |
Polyclonal | |
Unconjugated | |
ELOA2 | |
110kDa; AA408125; El; EloA; EloA2; Elongin 110 kDa subunit; elongin A; Elongin A2; elongin-A; Elongin-A2; MSTP059; RNA polymerase II transcription factor elongin subunit A (110 kDa); RNA polymerase II transcription factor SIII subunit A; RNA polymerase II transcription factor SIII subunit A1; SIII; SIII p110; TCEB3; TCEB3A; TCEB3B; TCEB3L; transcription elongation factor B (SIII), polypeptide 3; transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A); transcription elongation factor B alpha subunit; transcription elongation factor B polypeptide 3; transcription elongation factor B subunit 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27224, 51224, 6924 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q14241, Q8CB77, Q8IYF1 | |
ELOA, ELOA2 | |
A synthesized peptide derived from human ELOA(Accession Q14241), corresponding to amino acid residues E135-H185. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction