Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELOA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101331 Supplier Invitrogen™ Supplier No. PA5101331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ELOA3.

The elongin (SIII) complex is composed of a transcriptionally active A subunit and two small regulatory subunits, B and C. This complex stimulates the transcription elongation rate of RNA polyermase II by suppressing time spent at template-encoded arresting sites along DNA. Elongin A3, also known as RNA polymerase II transcription factor SIII subunit A3 and Transcription elongation factor B polypeptide 3C, is a 546 amino acid nuclear protein that functions as a general transcription elongation factor that increases RNA polymerase II transcription elongation and forms a stable complex with Elongin BC. In contrast to Elongin A, transcriptional activity of Elongin A3 is not stimulated by Elongin BC. Elongin A3 contains one TFIIS N-terminal domain and is ubiquitously expressed in human tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOA3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ELOA3
Gene Accession No. Q8NG57
Gene Alias eloA3; Elongin A3; ElonginA3; Elongin-A3; HsT829; RNA polymerase II transcription factor SIII subunit A3; TCEB3C; TCEB3L2; Transcription elongation factor B polypeptide 3C; transcription elongation factor B polypeptide 3C (elongin A3); transcription elongation factor B subunit 3C
Host Species Rabbit
Immunogen A synthesized peptide derived from human ELOA3(Accession Q8NG57), corresponding to amino acid residues A383-K433.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162699
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.