Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELOF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568347 Supplier Invitrogen™ Supplier No. PA568347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Eukaryotic RNA polymerase II mediates the synthesis of mature and functional messenger RNA. This is a multistep process, called the transcription cycle, that includes five stages: preinitiation, promoter, clearance, elongation and termination. Elongation is thought to be a critical stage for the regulation of gene expression. ELOF1 (Transcription elongation factor 1 homolog) is a 83 amino acid nuclear protein that is implicated in the maintenance of proper chromatin structure in regions that are being actively transcribed. The gene encoding ELOF1 maps to human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin (Ig) superfamily members, including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG family and Fc receptors (FcRs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOF1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.4
Gene ELOF1
Gene Accession No. P60002, P60003
Gene Alias 1110011K10Rik; ELF1; ELF1 homolog, elongation factor 1; ELOF1; elongation factor 1 homolog; elongation factor 1 homolog (ELF1, S. cerevisiae); elongation factor 1 homolog (S. cerevisiae); RGD1309855; transcription elongation factor 1 homolog
Gene Symbols ELOF1
Host Species Rabbit
Immunogen Synthesized peptide derived from human ELOF1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66126, 691193, 84337
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.