Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELOVL1 Rabbit anti-Human, Mouse, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502206871
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6871 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6871 Supplier Fabgennix Inc Supplier No. ELOVL1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ELOVL1 Polyclonal antibody specifically detects ELOVL1 in Human, Mouse, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ELOVL1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ELOVL1
Gene Accession No. Q9BW60, Q9JLJ5
Gene Alias 3-keto acyl-CoA synthase ELOVL1; AA407424; BB151133; CGI88; CGI-88; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 1; elongation of very long chain fatty acids protein 1; ELOVL FA elongase 1; ELOVL fatty acid elongase 1; ELOVL1; sequence similarity to Cig30 1; Ssc1; Very long chain 3-ketoacyl-CoA synthase 1; very long chain 3-oxoacyl-CoA synthase 1; very-long-chain 3-oxoacyl-CoA synthase 1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 148-198 on human Elongation of very long chain fatty acids protein 1.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 456820, 54325, 64834
Target Species Human, Mouse, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.