Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELOVL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120576 Supplier Invitrogen™ Supplier No. PA5120576
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF7, HepG2, Rat testis. Immunogen sequence: ILFLNFYVQT YRKKPMKKDM QEPPAGKEVK NGFSKAYFTA ANGVMNKKAQ.

ELOVL2 could be implicated in tissue-specific synthesis of very long chain fatty acids and sphingolipids. This protein may catalyze one or both of the reduction reaction in fatty acid elongation, i. e., conversion of beta-ketoacyl CoA to beta-hydroxyacyl CoA or reduction of trans-2-enoyl CoA to the saturated acyl CoA derivative.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOVL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ELOVL2
Gene Accession No. D4A612, Q9NXB9
Gene Alias 3-keto acyl-CoA synthase ELOVL2; AI317360; ELG3; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 2; elongation of very long chain fatty acids protein 2; elongation of very long chain fatty acids-like 2; ELOVL FA elongase 2; ELOVL family member 2; ELOVL fatty acid elongase 2; Elovl2; sequence similarity to Cig30 2; SSC2; Very long chain 3-ketoacyl-CoA synthase 2; very long chain 3-oxoacyl-CoA synthase 2; very-long-chain 3-oxoacyl-CoA synthase 2
Gene Symbols ELOVL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 247-296 of human ELOVL2 (NP_060240.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498728, 54898
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.