Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOVL3 Antibody (V61 P2B3*D10), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250385JF549
Description
ELOVL3 Monoclonal specifically detects ELOVL3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
ELOVL3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ELOVL3 | |
Ovalbumin-conjugated synthetic peptide - QPYNFELSK | |
0.1 mL | |
83401 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
V61 P2B3*D10 | |
Western Blot, ELISA, Immunohistochemistry | |
3-keto acyl-CoA synthase ELOVL3, CIG-30, CIG30MGC21435, Cold-inducible glycoprotein of 30 kDa, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3, elongation of very long chain fatty acids protein 3 | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction