Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOVL3 Antibody (V61P2B3-D10), Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $560.50
Specifications
Antigen | ELOVL3 |
---|---|
Clone | V61P2B3-D10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
ELOVL3 Monoclonal specifically detects ELOVL3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
ELOVL3 | |
1 mg/mL | |
Monoclonal | |
Mouse | |
PBS pH 7.4, 50% glycerol with 0.09% sodium azide | |
83401 | |
Synthetic peptide of Human Elongation of very long chain fatty acids protein 3 | |
Primary | |
Detects ∼40 kDa |
V61P2B3-D10 | |
Western Blot 1:1000, ELISA, Immunohistochemistry | |
Unconjugated | |
Human, Rat | |
3-keto acyl-CoA synthase ELOVL3, CIG-30, CIG30MGC21435, Cold-inducible glycoprotein of 30 kDa, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3, elongation of very long chain fatty acids protein 3 | |
ELOVL3 | |
IgG1 | |
Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title