Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELOVL3 Monoclonal Antibody (V61P2B3-D10), PerCP
GREENER_CHOICE

Catalog No. PIMA545908
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545908 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545908 Supplier Invitrogen™ Supplier No. MA545908
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45908 was sufficient for detection of Elongation of very long chain fatty acids protein 3 in 10 μg of Rat Brain by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects 40 kDa.

This gene encodes a protein that belongs to the GNS1/SUR4 family. Members of this family play a role in elongation of long chain fatty acids to provide precursors for synthesis of sphingolipids and ceramides. [provided by RefSeq, Jul 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOVL3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone V61P2B3-D10
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene ELOVL3
Gene Accession No. Q9HB03
Gene Alias 3-keto acyl-CoA synthase ELOVL3; CIG30; CIG-30; Cold-inducible glycoprotein of 30 kDa; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3; elongation of very long chain fatty acids protein 3; ELOVL FA elongase 3; ELOVL fatty acid elongase 3; ELOVL3; Very long chain 3-ketoacyl-CoA synthase 3; very long chain 3-oxoacyl-CoA synthase 3; very-long-chain 3-oxoacyl-CoA synthase 3
Gene Symbols ELOVL3
Host Species Mouse
Immunogen Synthetic peptide of Human Elongation of very long chain fatty acids protein 3.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309449, 83401
Target Species Human, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.