Learn More
Invitrogen™ ELOVL4 Polyclonal Antibody

Description
A suggested positive control for this product is brain tissue lysate.
Specifications
Specifications
| Antigen | ELOVL4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | ELOVL4 |
| Gene Accession No. | Q9GZR5 |
| Gene Alias | 3-keto acyl-CoA synthase Elovl4; ADMD; cancer/testis antigen 118; CT118; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4; elongation of very long chain fatty acids protein 4; elongation of very long chain fatty acids-like 4; ELOVL FA elongase 4; ELOVL fatty acid elongase 4; Elovl4; FLJ17667; FLJ92876; ISQMR; SCA34; STGD2; STGD3; Very long chain 3-ketoacyl-CoA synthase 4; very long chain 3-oxoacyl-CoA synthase 4; very-long-chain 3-oxoacyl-CoA synthase 4 |
| Gene Symbols | ELOVL4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.