Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOVL4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31192025UG
Description
ELOVL4 Polyclonal antibody specifically detects ELOVL4 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ELOVL4 | |
Polyclonal | |
Western Blot 1:400, Immunohistochemistry 10 - 15 ug/ml | |
ADMD, cancer/testis antigen 118, CT118, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4, elongation of very long chain fatty acids protein 4, FLJ17667, FLJ92876,3-keto acyl-CoA synthase ELOVL4, Stargardt disease 3 (autosomal dominant), STGD2, STGD3 | |
Synthetic peptdide derived from the human ELOVL4 protein (Uniprot: Q9GZR5) | |
25 μg | |
Lipid and Metabolism, Vision | |
6785 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction