Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOVL5 Antibody (Z88) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254809
Description
ELOVL5 Monoclonal specifically detects ELOVL5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
ELOVL5 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1, ELISA 1:1, Immunohistochemistry 1:1 | |
dJ483K16.1, EC 2.3.1.n8, elongation of very long chain fatty acids protein 5, ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, ELOVL2,3-keto acyl-CoA synthase ELOVL5, Fatty acid elongase 1, hELO1, HELO1homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2, homolog of yeast long chain polyunsaturated fatty acid elongatio, SUR4/Elo3-like, yeast) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Z88 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
ELOVL5 | |
Peptide Sequence NNVKPRKLR | |
0.1 mg | |
Cardiovascular Biology, Cellular Signaling | |
60481 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction