Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOVL7 Antibody (Z26), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250384AF750
Description
ELOVL7 Monoclonal specifically detects ELOVL7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
ELOVL7 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ELOVL7 | |
Peptide Sequence - RAYTKGQRLP (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), bovine (Bos taurus), canine (Canis familiaris), equine (Equus caballus) | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Z26 | |
Western Blot, ELISA, Immunohistochemistry | |
ELOVL family member 7, elongation of long chain fatty acids, ELOVL fatty acid elongase 7 | |
Mouse | |
Protein A purified | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
79993 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction