Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELOVL7 Antibody (Z26) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2503840.025ML
Description
ELOVL7 Monoclonal antibody specifically detects ELOVL7 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
ELOVL7 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
ELOVL family member 7, elongation of long chain fatty acids, ELOVL fatty acid elongase 7 | |
Peptide Sequence - RAYTKGQRLP (peptide immunogen is identical in rat (Rattus norvegicus), mouse (Mus musculus), bovine (Bos taurus), canine (Canis familiaris), equine (Equus caballus) | |
0.025 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
79993 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Z26 | |
Unconjugated | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction