Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELOVL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557615 Supplier Invitrogen™ Supplier No. PA557615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAFSDLTSRT VHLYDNWIKD ADPRVEDWLL M Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 48%.

Lipogenesis is a key event in the energy storage system and is controlled by the transcription factor sterol regulatory element-binding protein (SREBP)-1. Elongation of very long chain fatty acids protein 7 (ELOVL7) is a member of fatty acyl-CoA elongase gene family that elongates saturated very-long-chain fatty acids (SVLFA, C20:0-) and has been suggested to be involved in prostate cancer growth through saturated long-chain fatty acid metabolism. The metabolic pathways of long-chain fatty acids play an important role in the maintenance of membrane lipid composition and the generation of cell signaling precursor molecules such as eicosanoids and sphingosine-1 phosphate. Overexpression of ELOVL7 results in lipid accumulation in differentiated adipocytes; its expression is regulated by the microRNA miR-219.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOVL7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELOVL7
Gene Accession No. A1L3X0
Gene Alias 3-keto acyl-CoA synthase Elovl7; 9130013K24Rik; AI840082; elongation of very long chain fatty acids protein 7; elongation of very long chain fatty acids-like 7; ELOVL FA elongase 7; ELOVL family member 7, elongation of long chain fatty acids; ELOVL family member 7, elongation of long chain fatty acids (yeast); ELOVL fatty acid elongase 7; ELOVL7; Very long chain 3-ketoacyl-CoA synthase 7; very long chain 3-oxoacyl-CoA synthase 7; very-long-chain 3-oxoacyl-CoA synthase 7
Gene Symbols ELOVL7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELOVL7. Recombinant protein control fragment (Product #RP-96918).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.