Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551791
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551791 Supplier Invitrogen™ Supplier No. PA551791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ECDSTDDCIE HNIGPCSSVL DVGGAVTAVS VCPVLHPSQR YVVAVGLECG KICLYTWKKT DQVPEINDWT HCVETSQSQS HTLAIRKLCW KNCSGKTEQK EAEGAEWLHF ASCGEDHTVK IHRVNKCA Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 72%.

ELP2 regulates the ligand-dependent activation of STAT3. The protein acts as subunit of the RNA polymerase II elongator complex, which is a histone acetyltransferase component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Elp2
Gene Accession No. Q6IA86
Gene Alias AU023723; elongation protein 2 homolog; elongator acetyltransferase complex subunit 2; elongator complex protein 2; elongator protein 2; ELP2; Epl2; SHINC-2; signal transducer and activator of transcription 3 interacting protein 1; signal transducer and activator of transcription interacting protein 1; STAT3-interacting protein 1; Statip1; StIP; StIP1
Gene Symbols Elp2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELP2. Recombinant protein control fragment (Product #RP-94770).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.