Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elp3 Antibody (SR2083), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32182350UL
Description
Elp3 Monoclonal antibody specifically detects Elp3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Elp3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
55140 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Immunofluorescence, Immunoprecipitation | |
SR2083 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
EC 2.3.1.48, elongation protein 3 homolog (S. cerevisiae), elongator complex protein 3, hELP3, KAT9FLJ10422 | |
A synthesized peptide derived from human Elp3 (Uniprot #: Q9H9T3) | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction