Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELP3 Monoclonal Antibody (8B4)
GREENER_CHOICE

Catalog No. PIMA549241
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549241 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549241 Supplier Invitrogen™ Supplier No. MA549241
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human U20S whole cell, human HepG2 whole cell, human U87 whole cell, rat lung tissue, rat PC-12 whole cell, mouse lung tissue, mouse NIH/3T3 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELP3
Applications Western Blot
Classification Monoclonal
Clone 8B4
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene ELP3
Gene Accession No. Q9CZX0, Q9H9T3
Gene Alias 2610507P14Rik; EC 2.3.1.48; elongation protein 3 homolog; elongator acetyltransferase complex subunit 3; elongator complex protein 3; ELP3; h KAT9; hELP3; KAT9; LOC100362548; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase
Gene Symbols ELP3
Host Species Mouse
Immunogen A synthetic peptide corresponding to a sequence of human KAT9/ELP3 (RDPTKFQHQGFGMLLMEEAERIAREEH).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100362548, 55140, 74195
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.