Learn More
Invitrogen™ ELP3 Monoclonal Antibody (GT11811)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527840
Description
Positive Control: HeLa, HeLa nuclear, 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (96%), Zebrafish (92%), Drosophila (88%), Yeast (84%), Xenopus laevis (92%), Chicken (91%), Rhesus Monkey (100%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.
Specifications
ELP3 | |
Monoclonal | |
2.35 mg/mL | |
PBS with no preservative | |
Q9H9T3 | |
ELP3 | |
Recombinant protein encompassing a sequence within the center region of human ELP3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
GT11811 | |
Unconjugated | |
ELP3 | |
2610507P14Rik; EC 2.3.1.48; elongation protein 3 homolog; elongator acetyltransferase complex subunit 3; elongator complex protein 3; ELP3; h KAT9; hELP3; KAT9; LOC100362548; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase | |
Mouse | |
Protein G | |
RUO | |
55140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.