Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ELP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555260
Description
Immunogen sequence: ELWKSGRYKS YSPSDLVELV ARILALVPPW TRVYRVQRDI PMPLVSSGVE HGNLRELALA RMKDLGIQCR DVRTREVGIQ EIHHKVRPYQ VELVRR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.
Specifications
ELP3 | |
Polyclonal | |
Unconjugated | |
ELP3 | |
2610507P14Rik; EC 2.3.1.48; elongation protein 3 homolog; elongator acetyltransferase complex subunit 3; elongator complex protein 3; ELP3; h KAT9; hELP3; KAT9; LOC100362548; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100362548, 55140, 74195 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9CZX0, Q9H9T3 | |
ELP3 | |
Recombinant protein corresponding to Human ELP3. Recombinant protein control fragment (Product #RP-93794). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction