Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELP3 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992992
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2992 10 μL
50-199-2991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2992 Supplier Active Motif Supplier No. 39950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.

Specifications

Antigen ELP3
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene ELP3
Gene Accession No. Q9H9T3
Gene Alias 2610507P14Rik; EC 2.3.1.48; elongation protein 3 homolog; elongator acetyltransferase complex subunit 3; Elongator complex protein 3; Elp3; h KAT9; hELP3; KAT9; LOC100362548; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase
Gene Symbols ELP3
Host Species Rabbit
Immunogen This ELP3 antibody was raised against a peptide within the N-terminal half of human ELP3.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55140
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.