Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELP3 Recombinant Rabbit Monoclonal Antibody (3R3U5)
GREENER_CHOICE

Catalog No. PIMA542582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542582 Supplier Invitrogen™ Supplier No. MA542582
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: MCF7, K-562, SH-SY5Y, Mouse testis, Mouse brain, Mouse kidney, Rat lung, Rat testis, Rat brain. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MRQKRKGDLS PAELMMLTIG DVIKQLIEAH EQGKDIDLNK VKTKTAAKYG LSAQPRLVDI IAAVPPQYRK VLMPKLKAKP IRTASGIAVV AVMCKPHRCP HISFTGNICV YCPGGPDSDF EYSTQSYTGY EPTSMRAIRA RYDPFLQTRH RIEQLKQLGH SVDKVEFIVM GGTFMALPEE YRDYFIRNLH DALSGHTSNN IYEAVKYSER SLTKCIGITI ETRPDYCMKR HLSDMLTYGC TRLEIGVQSV YEDVARDTNR GHTVKAVCES FHLAKDSGFK VVAHMMPDLP NVGLERDIEQ FTEFFENPAF RPDGLKLYPT LVIRGTGLYE LWKSGRYKSY SPSDLVELVA RILALVPPWT RVYRVQRDIP MPLVSSGVEH GNLRELALAR MKDLGIQCRD VRTREVGIQE IHHKVRPYQV ELVRRDYVAN GGWETFLSYE DPDQDILIGL LRLRKCSEET FRFELGGGVS IVRELHVYGS VVPVSSRDPT KFQHQGFGML LMEEAERIAR EEHGSGKIAV ISGVGTRNYY RKIGYRLQGP YMVKMLK.

Elongator complex protein 3 (ELP3) is a catalytic histone acetyltransferase subunit of the RNA polymerase II elongator complex, which is a component of the RNA polymerase II (Pol II) holoenzyme and is involved in transcriptional elongation. Elongator may play a role in chromatin remodeling and is involved in acetylation of histones H3 and probably H4. It may also have a methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 3R3U5
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ELP3
Gene Accession No. Q9CZX0, Q9H9T3
Gene Alias 2610507P14Rik; EC 2.3.1.48; elongation protein 3 homolog; elongator acetyltransferase complex subunit 3; elongator complex protein 3; ELP3; h KAT9; hELP3; KAT9; LOC100362548; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase
Gene Symbols ELP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-547 of human KAT9/Elp3 (Q9H9T3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100362548, 55140, 74195
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.