Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elp4 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212785V
Description
Elp4 Polyclonal antibody specifically detects Elp4 in Human samples. It is validated for Western BlotSpecifications
Elp4 | |
Polyclonal | |
Western Blot | |
C11orf19, chromosome 11 open reading frame 19, dJ68P15A.1, elongation protein 4 homolog (S. cerevisiae), elongator complex protein 4, hELP4, PAX6 neighbor gene protein, PAX6NEB, PAXNEBFLJ20498 | |
The immunogen this antibody was made to, maps to a region between residue 374 and 424 of human Elongation Protein 4 using the numbering given in entry NP_061913.3 (GeneID 26610). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
26610 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction