Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elp4 Antibody (SR1794), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32247850UL
Description
Elp4 Monoclonal antibody specifically detects Elp4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Elp4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1794 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
C11orf19, chromosome 11 open reading frame 19, dJ68P15A.1, elongation protein 4 homolog (S. cerevisiae), elongator complex protein 4, hELP4, PAX6 neighbor gene protein, PAX6NEB, PAXNEBFLJ20498 | |
A synthesized peptide derived from human Elp4 (Uniprot #: Q96EB1) | |
50 μg | |
DNA replication Transcription Translation and Splicing | |
26610 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction