Learn More
Invitrogen™ ELP4 Monoclonal Antibody (OTI4H10)

Description
ELP4 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ELP4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4H10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Elp4 |
| Gene Accession No. | Q96EB1 |
| Gene Alias | A330107A17Rik; AN; C11orf19; dJ68P15A.1; elongation protein 4 homolog; elongation protein 4 homolog (S. cerevisiae); elongator acetyltransferase complex subunit 4; elongator complex protein 4; ELP4; hELP4; hypothetical protein LOC614139; PAX6 neighbor gene protein; PAX6NEB; PAXNEB; RGD1566052; similar to elongation protein 4 homolog |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.