Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568850
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568850 Supplier Invitrogen™ Supplier No. PA568850
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 79%; Horse: 91%; Human: 79%; Mouse: 100%; Pig: 79%; Rabbit: 79%; Rat: 90%.

This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELP4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Elp4
Gene Accession No. Q9ER73
Gene Alias A330107A17Rik; AN; C11orf19; dJ68P15A.1; elongation protein 4 homolog; elongation protein 4 homolog (S. cerevisiae); elongator acetyltransferase complex subunit 4; elongator complex protein 4; ELP4; hELP4; hypothetical protein LOC614139; PAX6 neighbor gene protein; PAX6NEB; PAXNEB; RGD1566052; similar to elongation protein 4 homolog
Gene Symbols Elp4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse ELP4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 77766
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.