Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Elplasia™ Microcavity Plates
GREENER_CHOICE

Catalog No. 07202464
Click to view available options
Well Count:
6
24
Surface Treatment:
Low Cell Bind
Tissue Culture

The only plates with a simple 'plug and play' protocol for scaffold-free, self-assembly spheroid formation at large volumes.

  • Novel well geometry and surface coatings, including Corning Ultra Low Attachment (ULA) featured on the round bottom plates
  • Corning ULA surface is hydrophilic, biologically inert and non-degradable, which promotes highly reproducible spheroid formation and easy harvesting
  • Corning Elplasia round bottom plates optimal for bulk spheroid formation, collection and expansion\
  • Round bottom plates feature Corning Ultra-Low Attachment surface and are available in 6-, 24- and 96-well formats
  • Corning Elplasia square bottom plates feature a surface with optical qualities suited for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters
  • The Elplasia square plate is well suited for screening programs as it generates a high volume of spheroids per well, allowing for higher signals per well
  • Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats
  • Spheroids may be formed and cultured for 21 or more days
  • Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
  • The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
  • Black opaque sidewalls to reduce well-to-well 'cross-talk'
  • Well suited for fluorescent/luminescent assays
  • Available in multiple formats, two well geometries and surface coating options
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Round
Color Black, Clear
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Tissue Culture
Type Microcavity Plate
Well Count 6
Includes Lid
Quantity 5
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.