Learn More
3M™ Scott™ ELSA™ Emergency Escape Breathing Device - N (EEBD-N)
ELSA™ EEBD-N respirator emergency escape breathing device is designed for simple use, training, and maintenance. Carry bag offers improved functionality while the redesigned hood fits wearers comfortably.
Supplier: 3M™ Scott™ 7012473801
Description
Enables workers to escape from certain toxic environments, even in atmospheres immediately dangerous to life or health (IDLH).
- Features one-size-fits-all clear hood with patent-pending neck seal
- Pulls on with no additional steps to secure, making it easy to train and use properly
- Includes fully-enclosed, chemical-resistant PVC carry bag to keep it free of insects and debris
- Available with 5, 10, or 15 minute durations
- Available with full or drained cylinders
- NIOSH approved
- 10 and 15 minute models meet SOLAS Chapter II-2, Part D requirements.
- Fits into standard wall cases, has indefinite shelf life, and comes with a 3 year warranty.
Specifications
| NIOSH | |
| 5 min. | |
| Clear hood with patent-pending neck seal fits all wearers, and pulls on with no additional steps to secure, making it easy to train and use properly. | |
| 8007212-D |
| Escape Breathing Apparatus | |
| 30 lpm | |
| Respirator, Hood, Carry Bag, Drained Cylinder |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.