Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Elsilimomab Recombinant Mouse Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541861
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541861 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541861 Supplier Invitrogen™ Supplier No. MA541861
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Elsilimomab is a biosimilar that targets IL6. This gene encodes a cytokine that functions in inflammation and the maturation of B cells. The protein is primarily produced at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elsilimomab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias B-E8
Host Species Mouse
Immunogen Human IL6.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.