Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ELSPBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576076
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ELSPBP1 (epididymal sperm-binding protein 1), also known as epididymal secretory protein 12 (E12), HE12 or EDDM12, is a 223 amino acid member of the seminal plasma protein family. ELSPBP1 is a secreted protein that is detected in cauda epididymal fluid and on sperm membrane. ELSPBP1 has phosphorylcholine-binding activity and has been shown to bind to spermatozoa upon ejaculation and is thought to play a role in sperm capacitation. N-glycosylated, ELSPBP1 contains four fibronectin type-II domains. The gene that encodes ELSPBP1 maps to human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.
Specifications
ELSPBP1 | |
Polyclonal | |
Unconjugated | |
ELSPBP1 | |
E12; EDDM12; EL149; ELSPBP1; epididymal protein 12; epididymal secretory protein 12; epididymal sperm binding protein 1; epididymal sperm-binding protein 1; epididymis luminal secretory protein 149; ESPB1; hE12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BH3 | |
ELSPBP1 | |
Recombinant full length Human ELSPBP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction