Learn More
Invitrogen™ ELTD1 Polyclonal Antibody

Description
PA1-4203 detects the ETL protein in human samples. PA1-4203 has been successfully used in Western blot procedures. The PA1-4203 immunogen is a synthetic peptide corresponding to residues R(7) S S S N Q D R F I T N D(19) of human ETL.
Specifications
Specifications
| Antigen | ELTD1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ADGRL4 |
| Gene Accession No. | Q9HBW9 |
| Gene Alias | 1110033N21Rik; ADGRL4; Adhesion G protein-coupled receptor L4; EGF, latrophilin and seven transmembrane domain containing 1; EGF, latrophilin and seven transmembrane domain-containing protein 1; EGF, latrophilin seven transmembrane domain containing 1; EGF, latrophilin seven transmembrane domain-containing protein 1; EGF,latrophilin and seven transmembrane domain-containing protein 1; EGF-TM7 receptor; EGF-TM7-latrophilin-related protein; Eltd1; ETL; ETL protein; ETL1; hCG_14667; KPG_003; UNQ202/PRO228 |
| Gene Symbols | ADGRL4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.