Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eluent Solution, for Ion Chromatography, 1.7 mM NaHCO3 - 1.8 mM Na2CO3, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
1 L
4 L
Specifications
Specifications
Chemical Name or Material | Eluent Solution |
Color | Colorless |
Name Note | 1.7mM NaHCO |
CAS | 7732-18-5, 497-19-8, 144-55-8 |
CAS Min % | 98.0, 0.02, 0.01 |
CAS Max % | 100.0, 0.02, 0.01 |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Quantity | 4 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction