Learn More
Thermo Scientific™ Elution Buffer for Pierce™ IgM Purification Kit PROMO
Description
Pierce MBP Agarose is purified mannan binding protein (also called mannose-binding lectin, MBL) that has been immobilized on 4% beaded agarose. When used with an optimized buffer system based on temperature- and calcium-dependent binding, this resin enables specific affinity purification of mouse IgM from ascites fluid and other concentrated samples. The resin and essential buffers are available individually or bundled together in a complete IgM Purification Kit containing a pre-packed 5 mL column of the MBP Agarose.
Highlights:
Immobilized MBP – affinity system based on mannan binding protein (MBP) allows specific purification of mouse IgM from ascites fluid or partial isolation of human IgM from serum
Complete kit – IgM Purification Kit includes a pre-packed MBP column and ready-to-use buffers for convenient purification of at least 1.5mg IgM from 0.5 mL ascites samples per column-use
Mild conditions – nondenaturing, near-neutral buffer conditions ensure recovery of functional antibodies
Reusable – stable ligand and resins can be regenerated and reused at least 10 times without significant decline in binding capacity
Includes:
Kit contains a 5 mL MBP Agarose Column, column preparation buffer, IgM binding and elution buffers, and a column extender
Specifications
Specifications
| Product Type | IgM Purification Kit |
| Product Line | Pierce™ |
| Format | Kit |
| Reagent Type | Elution Buffer |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2°C to 8°C. |
| Quantity | 500 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.