Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELYS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$416.50 - $682.00
Specifications
Antigen | ELYS |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP) |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Proximity Ligation Assay |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ELYS Polyclonal specifically detects ELYS in Human, Bovine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Proximity Ligation Assay.Specifications
ELYS | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Proximity Ligation Assay | |
Unconjugated | |
RUO | |
Human, Bovine | |
AT hook containing transcription factor 1, ELYS, ELYS transcription factor like protein TMBS62, ELYS transcription factor-like protein TMBS62, MST108, MSTP108, protein ELYS, putative AT-hook-containing transcription factor 1, TMBS62 | |
AHCTF1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP) | |
Polyclonal | |
Rabbit | |
Transcription Factors and Regulators | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
25909 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GEEARNLSFNELYPSGTLKLQYNFDTIDQQFCDLADNKDTAECDIAEVDGELFVAQSNFTLILEGEEGEVEPGDFASSDVLPKAANTATEEKLVCSGEND | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title