Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EM 12-FS
GREENER_CHOICESDP

Catalog No. 785150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-515-0 50 mg
78-511-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 78-515-0 Supplier Tocris Bioscience™ Supplier No. 7851/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Molecular glue; induces degradation of NQAT1

EM12-FS is a selective N-terminal glutamine amidohydrolase 1 (NTAQ1) Degrader. In MOLT4 cells, it downregulates NTAQ1 via cereblon-mediated degradation (IC50 = 256 nM). Requires covalent His353 engagement to recruit and degrade NTAQ1.

Specifications

Chemical Name or Material 3-[6-[(Fluorosulfonyl)oxy]-1,3-dihydro-1-oxo-2H-isoindol-2-yl]-2,6-piperidinedione
CAS 2839138-44-0
Quantity 50 mg
Target Molecular Glues
Molecular Formula C13H11FN2O6S
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.