Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMAP-II/AIMP1 Antibody (959507), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $499.50
Specifications
Antigen | EMAP-II/AIMP1 |
---|---|
Clone | 959507 |
Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB19101SP
![]() |
Novus Biologicals
MAB19101SP |
25 μg |
Each for $243.50
|
|
|||||
MAB19101
![]() |
Novus Biologicals
MAB19101 |
100 μg |
Each for $499.50
|
|
|||||
Description
EMAP-II/AIMP1 Monoclonal specifically detects EMAP-II/AIMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
EMAP-II/AIMP1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
9255 | |
IgG2b | |
Protein A or G purified |
959507 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43 | |
E.coli-derived recombinant human EMAP-II Ser147-Lys312 Accession # Q12904 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title