Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMAP-II/AIMP1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298343AF405
Description
EMAP-II/AIMP1 Polyclonal specifically detects EMAP-II/AIMP1 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
EMAP-II/AIMP1 | |
Polyclonal | |
Western Blot, ELISA | |
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43 | |
Produced in rabbits immunized with purified, recombinant EMAP-II/AIMP1 (Ser145-Lys310). | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
9255 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction